Assay Details
Gene Symbol: BABAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005781.3 | XP_004005830.1 | 4 - 5 | 433 | 84 | |
XM_012166941.2 | XP_012022331.1 | 5 - 6 | 525 | 84 | ||
XM_027966635.1 | XP_027822436.1 | 4 - 5 | 348 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
