Assay Details
Gene Symbol: BICRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978541.1 | XP_027834342.1 | 12 - 13 | 3661 | 109 | |
XM_027978542.1 | XP_027834343.1 | 12 - 13 | 3742 | 109 |

More Information
Set Membership: |
![]() ![]() |
