Assay Details
Gene Symbol: LOC105607297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971828.1 | XP_027827629.1 | 4 - 5 | 1780 | 68 | |
XM_027971829.1 | XP_027827630.1 | 4 - 5 | 1780 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
