Assay Details
Gene Symbol: FAM155A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184938.3 | XP_012040328.2 | 2 - 3 | 1340 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM155A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184938.3 | XP_012040328.2 | 2 - 3 | 1340 | 69 |
Set Membership: |
![]() ![]() ![]() |