Assay Details
Gene Symbol: LOC114118509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979878.1 | XP_027835679.1 | 4 - 5 | 317 | 66 |
Gene Symbol: LOC101110537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979879.1 | XP_027835680.1 | 7 - 8 | 716 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
