Assay Details
Gene Symbol: LOC101103343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015280.4 | XP_004015329.1 | 7 - 8 | 1338 | 86 | |
XM_027978377.1 | XP_027834178.1 | 7 - 8 | 1227 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
