Assay Details
Gene Symbol: LOC101106395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004155.4 | XP_004004204.1 | 11 - 12 | 1920 | 99 | |
XM_027964301.1 | XP_027820102.1 | 12 - 13 | 2046 | 99 | ||
XM_027964302.1 | XP_027820103.1 | 11 - 12 | 1969 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
