Assay Details
Gene Symbol: PTCHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018873.4 | XP_004018922.2 | 1 - 2 | 425 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
