Assay Details
Gene Symbol: LOC114117326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976155.1 | XP_027831956.1 | 2 | 876 | 91 |
Gene Symbol: BOD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971185.1 | XP_027826986.1 | 15 - 16 | 8094 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
