Assay Details
Gene Symbol: ASTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103531.3 | XP_011958921.1 | 3 - 4 | 1894 | 93 | |
XM_012103575.3 | XP_011958965.1 | 3 - 4 | 1894 | 93 | ||
XM_027960594.1 | XP_027816395.1 | 3 - 4 | 1655 | 93 | ||
XM_027960603.1 | XP_027816404.1 | 2 - 3 | 1722 | 93 | ||
XM_027960605.1 | XP_027816406.1 | 3 - 4 | 1894 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
