Assay Details
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017912.4 | XP_004017961.2 | 22 - 23 | 2038 | 86 | |
XM_012098847.3 | XP_011954237.2 | 28 - 29 | 3861 | 86 | ||
XM_012098849.3 | XP_011954239.2 | 27 - 28 | 3834 | 86 | ||
XM_012098850.3 | XP_011954240.2 | 27 - 28 | 3732 | 86 | ||
XM_012098851.3 | XP_011954241.2 | 26 - 27 | 3705 | 86 | ||
XM_012098852.3 | XP_011954242.2 | 23 - 24 | 1987 | 86 | ||
XM_012098853.3 | XP_011954243.2 | 23 - 24 | 3055 | 86 | ||
XM_012098854.3 | XP_011954244.2 | 23 - 24 | 2065 | 86 | ||
XM_012098855.3 | XP_011954245.2 | 23 - 24 | 2935 | 86 | ||
XM_012098857.3 | XP_011954247.2 | 23 - 24 | 2926 | 86 | ||
XM_012098858.3 | XP_011954248.2 | 21 - 22 | 1909 | 86 | ||
XM_027957293.1 | XP_027813094.1 | 22 - 23 | 1936 | 86 | ||
XM_027957294.1 | XP_027813095.1 | 22 - 23 | 2925 | 86 | ||
XM_027957295.1 | XP_027813096.1 | 22 - 23 | 2797 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
