Assay Details
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970991.1 | XP_027826792.1 | 6 - 7 | 542 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970991.1 | XP_027826792.1 | 6 - 7 | 542 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |