Assay Details
Gene Symbol: ECPAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092979.2 | XP_014948465.2 | 42 - 43 | 4658 | 114 | |
XM_015092980.2 | XP_014948466.2 | 41 - 42 | 4581 | 114 | ||
XM_027964118.1 | XP_027819919.1 | 41 - 42 | 4841 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
