Assay Details
Gene Symbol: NFATC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961072.1 | XP_027816873.1 | 4 - 5 | 1922 | 70 | |
XM_027961073.1 | XP_027816874.1 | 4 - 5 | 1708 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
