Assay Details
Gene Symbol: LOC101122940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012175443.3 | XP_012030833.2 | 24 - 25 | 3105 | 74 | |
XM_012175444.3 | XP_012030834.2 | 24 - 25 | 3325 | 74 | ||
XM_012175446.3 | XP_012030836.2 | 24 - 25 | 3312 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
