Assay Details
Gene Symbol: PRDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001918.3 | XP_004001967.1 | 2 - 3 | 198 | 90 | |
XM_012126652.3 | XP_011982042.2 | 2 - 3 | 178 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
