Assay Details
Gene Symbol: PLA2G1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017414.4 | XP_004017463.1 | 3 - 4 | 438 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PLA2G1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017414.4 | XP_004017463.1 | 3 - 4 | 438 | 88 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |