Assay Details
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964603.1 | XP_027820404.1 | 4 - 5 | 1079 | 68 | |
XM_027964604.1 | XP_027820405.1 | 3 - 4 | 541 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
