Assay Details
Gene Symbol: LOC101116707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964869.1 | XP_027820670.1 | 15 - 16 | 1448 | 96 | |
XM_027964870.1 | XP_027820671.1 | 15 - 16 | 1480 | 96 | ||
XM_027964871.1 | XP_027820672.1 | 14 - 15 | 1352 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
