Assay Details
Gene Symbol: MYCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973699.1 | XP_027829500.1 | 24 - 25 | 3845 | 83 |
Gene Symbol: LOC114116662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973851.1 | XP_027829652.1 | 6 - 7 | 721 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
