Assay Details
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979522.1 | XP_027835323.1 | 9 - 10 | 1642 | 57 | |
XM_027979523.1 | XP_027835324.1 | 9 - 10 | 1225 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
