Assay Details
Gene Symbol: GPSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100905.3 | XP_011956295.1 | 1 - 2 | 434 | 67 | |
XM_015102774.2 | XP_014958260.1 | 1 - 2 | 425 | 67 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GPSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100905.3 | XP_011956295.1 | 1 - 2 | 434 | 67 | |
XM_015102774.2 | XP_014958260.1 | 1 - 2 | 425 | 67 |