Assay Details
Gene Symbol: PLPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104607.2 | XP_014960093.2 | 5 - 6 | 495 | 70 | |
XM_027962718.1 | XP_027818519.1 | 5 - 6 | 528 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
