Assay Details
Gene Symbol: LOC114112156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963732.1 | XP_027819533.1 | 6 - 7 | 1669 | 76 |
Gene Symbol: DHX57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966915.1 | XP_027822716.1 | 8 - 9 | 1961 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
