Assay Details
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094896.2 | XP_014950382.1 | 2 - 3 | 261 | 109 | |
XM_027967804.1 | XP_027823605.1 | 2 - 3 | 184 | 109 | ||
XM_027967805.1 | XP_027823606.1 | 2 - 3 | 297 | 109 | ||
XM_027967806.1 | XP_027823607.1 | 2 - 3 | 298 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
