Assay Details
Gene Symbol: TENT4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980174.1 | XP_027835975.1 | 10 - 11 | 1886 | 86 | |
XM_027980175.1 | XP_027835976.1 | 10 - 11 | 1886 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
