Assay Details
Gene Symbol: HDHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968063.1 | XP_027823864.1 | 2 - 3 | 317 | 83 | |
XM_027968064.1 | XP_027823865.1 | 2 - 3 | 410 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
