Assay Details
Gene Symbol: LOC114115233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970740.1 | XP_027826541.1 | 1 - 2 | 79 | 101 |
Gene Symbol: LOC101102485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970981.1 | XP_027826782.1 | 8 - 9 | 673 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
