Assay Details
Gene Symbol: LOC101115486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012098423.3 | XP_011953813.2 | 32 - 33 | 7377 | 59 | |
XM_012098428.3 | XP_011953818.2 | 27 - 28 | 5361 | 59 | ||
XM_027975368.1 | XP_027831169.1 | 36 - 37 | 8084 | 59 | ||
XM_027975369.1 | XP_027831170.1 | 36 - 37 | 8081 | 59 | ||
XM_027975370.1 | XP_027831171.1 | 36 - 37 | 8081 | 59 | ||
XM_027975371.1 | XP_027831172.1 | 36 - 37 | 8078 | 59 | ||
XM_027975372.1 | XP_027831173.1 | 35 - 36 | 8018 | 59 | ||
XM_027975373.1 | XP_027831174.1 | 34 - 35 | 7964 | 59 |
Gene Symbol: LOC114109118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957513.1 | XP_027813314.1 | 2 - 3 | 593 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
