Assay Details
Gene Symbol: LOC101115486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012098423.3 | XP_011953813.2 | 33 - 34 | 7713 | 82 | |
XM_012098428.3 | XP_011953818.2 | 28 - 29 | 5697 | 82 | ||
XM_027975368.1 | XP_027831169.1 | 37 - 38 | 8420 | 82 | ||
XM_027975369.1 | XP_027831170.1 | 37 - 38 | 8417 | 82 | ||
XM_027975370.1 | XP_027831171.1 | 37 - 38 | 8417 | 82 | ||
XM_027975371.1 | XP_027831172.1 | 37 - 38 | 8414 | 82 | ||
XM_027975372.1 | XP_027831173.1 | 36 - 37 | 8354 | 82 | ||
XM_027975373.1 | XP_027831174.1 | 35 - 36 | 8300 | 82 |
Gene Symbol: LOC114109118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957513.1 | XP_027813314.1 | 3 - 4 | 935 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
