Assay Details
Gene Symbol: CRYBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972398.1 | XP_027828199.1 | 13 - 14 | 5345 | 95 | |
XM_027972399.1 | XP_027828200.1 | 12 - 13 | 5257 | 95 | ||
XM_027972400.1 | XP_027828201.1 | 12 - 13 | 5058 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
