Assay Details
Gene Symbol: NT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098961.2 | XP_014954447.2 | 4 - 5 | 537 | 105 | |
XM_027974059.1 | XP_027829860.1 | 4 - 5 | 635 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
