Assay Details
Gene Symbol: DYNC2I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969086.1 | XP_027824887.1 | 9 - 10 | 1168 | 74 | |
XM_027969087.1 | XP_027824888.1 | 12 - 13 | 1625 | 74 | ||
XM_027969088.1 | XP_027824889.1 | 9 - 10 | 1427 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
