Assay Details
Gene Symbol: NAXE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002621.3 | XP_004002670.2 | 3 - 4 | 442 | 75 | |
XM_012183404.2 | XP_012038794.2 | 3 - 4 | 429 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
