Assay Details
Gene Symbol: NAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017115.3 | XP_004017164.2 | 9 - 10 | 1230 | 106 | |
XM_012096791.2 | XP_011952181.2 | 9 - 10 | 1230 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
