Assay Details
Gene Symbol: BICRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978541.1 | XP_027834342.1 | 10 - 11 | 3454 | 69 | |
XM_027978542.1 | XP_027834343.1 | 10 - 11 | 3535 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
