Assay Details
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092234.2 | XP_014947720.2 | 1 - 2 | 1387 | 101 | |
GenBank mRNA | AY535402.1 | - | 1 - 2 | 163 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
