Assay Details
Gene Symbol: LOC101105130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012097121.3 | XP_011952511.2 | 4 - 5 | 413 | 81 |
Gene Symbol: LOC114113818 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965986.1 | XP_027821787.1 | 4 - 5 | 413 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
