Assay Details
Gene Symbol: DOHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008617.4 | XP_004008666.2 | 3 - 4 | 739 | 112 | |
XM_027969887.1 | XP_027825688.1 | 3 - 4 | 709 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
