Assay Details
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002786.3 | XP_004002835.1 | 3 - 4 | 544 | 90 | |
XM_012186695.2 | XP_012042085.1 | 4 - 5 | 295 | 90 | ||
XM_012186700.2 | XP_012042090.1 | 3 - 4 | 273 | 90 | ||
XM_012186706.3 | XP_012042096.1 | 3 - 4 | 186 | 90 | ||
XM_012186712.2 | XP_012042102.1 | 4 - 5 | 600 | 90 | ||
XM_027978512.1 | XP_027834313.1 | 4 - 5 | 327 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
