Assay Details
Gene Symbol: SPACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012160807.3 | XP_012016197.2 | 3 - 4 | 482 | 84 | |
XM_012160825.3 | XP_012016215.2 | 2 - 3 | 289 | 84 | ||
XM_027966234.1 | XP_027822035.1 | 2 - 3 | 1359 | 84 | ||
XM_027966235.1 | XP_027822036.1 | 2 - 3 | 289 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
