Assay Details
Gene Symbol: LOC101108212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967845.1 | XP_027823646.1 | 1 - 2 | 759 | 83 |
Gene Symbol: LOC114114054 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967844.1 | XP_027823645.1 | 1 - 2 | 163 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
