Assay Details
Gene Symbol: LOC101105324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008102.3 | XP_004008151.2 | 5 - 6 | 817 | 122 | |
XM_012177227.3 | XP_012032617.1 | 6 - 7 | 891 | 122 | ||
XM_012177228.3 | XP_012032618.1 | 6 - 7 | 631 | 122 | ||
XM_027968861.1 | XP_027824662.1 | 5 - 6 | 801 | 122 | ||
XM_027968862.1 | XP_027824663.1 | 4 - 5 | 424 | 122 | ||
XM_027968863.1 | XP_027824664.1 | 6 - 7 | 1150 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
