Assay Details
Gene Symbol: PLPPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970142.1 | XP_027825943.1 | 5 - 6 | 677 | 81 | |
XM_027970143.1 | XP_027825944.1 | 5 - 6 | 696 | 81 | ||
XM_027970144.1 | XP_027825945.1 | 5 - 6 | 677 | 81 | ||
XM_027970145.1 | XP_027825946.1 | 4 - 5 | 1517 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
