Assay Details
Gene Symbol: PPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104411.2 | XP_014959897.2 | 9 - 10 | 958 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
