Assay Details
Gene Symbol: ECPAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092979.2 | XP_014948465.2 | 4 - 5 | 391 | 95 | |
XM_015092980.2 | XP_014948466.2 | 3 - 4 | 314 | 95 | ||
XM_027964118.1 | XP_027819919.1 | 3 - 4 | 574 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
