Assay Details
Gene Symbol: LOC101114342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006170.4 | XP_004006219.1 | 6 - 7 | 957 | 104 | |
XM_012173857.3 | XP_012029247.1 | 7 - 8 | 1001 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
