Assay Details
Gene Symbol: ERRFI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099237.2 | XP_014954723.2 | 2 - 3 | 5415 | 102 | |
XM_027975677.1 | XP_027831478.1 | 3 - 4 | 411 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
