Assay Details
Gene Symbol: ARL6IP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956439.1 | XP_027812240.1 | 3 - 4 | 835 | 79 | |
XM_027956440.1 | XP_027812241.1 | 3 - 4 | 811 | 79 | ||
XM_027956441.1 | XP_027812242.1 | 3 - 4 | 835 | 79 | ||
XM_027956442.1 | XP_027812243.1 | 3 - 4 | 1080 | 79 | ||
XM_027956443.1 | XP_027812244.1 | 4 - 5 | 743 | 79 | ||
XM_027956444.1 | XP_027812245.1 | 4 - 5 | 716 | 79 | ||
XM_027956445.1 | XP_027812246.1 | 4 - 5 | 713 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
