Assay Details
Gene Symbol: LOC101120595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977130.1 | XP_027832931.1 | 24 - 25 | 3042 | 64 | |
XM_027977131.1 | XP_027832932.1 | 24 - 25 | 3042 | 64 | ||
XM_027977132.1 | XP_027832933.1 | 24 - 25 | 3042 | 64 | ||
XM_027977133.1 | XP_027832934.1 | 24 - 25 | 3042 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
